Assay Details
Gene Symbol: GPR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973461.1 | XP_027829262.1 | 1 | 1808 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
