Assay Details
| Gene Symbol: LOC101111473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187510.2 | XP_012042900.2 | 3 | 1321 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111473| Gene Symbol: LOC101111473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187510.2 | XP_012042900.2 | 3 | 1321 | 98 | |
Set Membership: |
|