Assay Details
| Gene Symbol: RBBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005013.3 | XP_004005062.1 | 12 | 1758 | 107 | |
| Gene Symbol: LOC106991292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962606.1 | XP_027818407.1 | 1 | 1717 | 107 | |
More Information
Set Membership: |
|

RBBP4
Genomic Map