Assay Details
| Gene Symbol: TP53INP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977111.1 | XP_027832912.1 | 3 | 498 | 76 | |
| XM_027977112.1 | XP_027832913.1 | 4 | 608 | 76 | ||
| XM_027977113.1 | XP_027832914.1 | 3 | 462 | 76 | ||
More Information
Set Membership: |
|

TP53INP2
Genomic Map