Assay Details
Gene Symbol: PAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978269.1 | XP_027834070.1 | 6 | 1385 | 97 | |
XM_027978270.1 | XP_027834071.1 | 6 | 1443 | 97 | ||
XM_027978271.1 | XP_027834072.1 | 6 | 1495 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
