Assay Details
| Gene Symbol: RAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104814.2 | XP_014960300.2 | 2 | 455 | 92 | |
| XM_027963023.1 | XP_027818824.1 | 2 | 312 | 92 | ||
More Information
Set Membership: |
|

RAI2
Genomic Map