Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960746.1 | XP_027816547.1 | 7 | 2909 | 102 | |
| XM_027960747.1 | XP_027816548.1 | 6 | 2803 | 102 | ||
More Information
Set Membership: |
|

RNF138
Genomic Map