Assay Details
Gene Symbol: MICOS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965599.1 | XP_027821400.1 | 4 | 502 | 101 | |
XM_027965600.1 | XP_027821401.1 | 4 | 512 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
