Assay Details
Gene Symbol: MIR541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107928.1 | 1 | 48 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|
