Assay Details
| Gene Symbol: CSNK1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082373.1 | NP_001075842.1 | 2 - 3 | 607 | 125 | |
| XM_008254851.2 | XP_008253073.1 | 2 - 3 | 818 | 125 | ||
| XM_008254854.2 | XP_008253076.1 | 2 - 3 | 817 | 125 | ||
| XM_008254855.2 | XP_008253077.1 | 2 - 3 | 811 | 125 | ||
| XM_008254856.2 | XP_008253078.1 | 2 - 3 | 814 | 125 | ||
| XM_008254857.2 | XP_008253079.1 | 2 - 3 | 818 | 125 | ||
| XM_008254858.2 | XP_008253080.1 | 2 - 3 | 812 | 125 | ||
| XM_008254859.2 | XP_008253081.1 | 2 - 3 | 801 | 125 | ||
| XM_008254860.2 | XP_008253082.1 | 2 - 3 | 805 | 125 | ||
| GenBank mRNA | U59166.1 | - | 2 - 3 | 607 | 125 | |
More Information
Set Membership: |
|

CSNK1A1
Genomic Map