Assay Details
| Gene Symbol: DDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722701.3 | XP_002722747.1 | 864 | 77 | ||
| XM_008250483.1 | XP_008248705.1 | 859 | 77 | |||
| XM_008250484.2 | XP_008248706.1 | 859 | 77 | |||
More Information
Set Membership: |
|

DDX6
Genomic Map