Assay Details
| Gene Symbol: VDAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082718.1 | NP_001076187.1 | 6 - 7 | 581 | 152 | |
| XM_008269908.2 | XP_008268130.1 | 7 - 8 | 751 | 152 | ||
| GenBank mRNA | AF209726.1 | - | 6 - 7 | 581 | 152 | |
More Information
Set Membership: |
|

VDAC2
Genomic Map