Assay Details
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204847.1 | NP_001191776.1 | 8 - 9 | 1395 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
