Assay Details
| Gene Symbol: DDX50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718449.3 | XP_002718495.1 | 10 - 11 | 1635 | 126 | |
| XM_008270034.2 | XP_008268256.1 | 11 - 12 | 1746 | 126 | ||
| XM_008270035.2 | XP_008268257.1 | 11 - 12 | 1688 | 126 | ||
| XM_017348734.1 | XP_017204223.1 | 10 - 11 | 1537 | 126 | ||
More Information
Set Membership: |
|

DDX50
Genomic Map