Assay Details
| Gene Symbol: DERL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724013.3 | XP_002724059.1 | 264 | 60 | ||
| XM_008252996.2 | XP_008251218.1 | 260 | 60 | |||
| XM_008252997.2 | XP_008251219.1 | 190 | 60 | |||
More Information
Set Membership: |
|

DERL3
Genomic Map