Assay Details
| Gene Symbol: LOC100008606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017340621.1 | XP_017196110.1 | 792 | 139 | ||
| GenBank mRNA | AB016250.1 | - | 365 | 139 | ||
| Gene Symbol: MX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723759.3 | XP_002723805.1 | 868 | 139 | ||
| XM_008252472.2 | XP_008250694.1 | 714 | 139 | |||
| XM_008252473.2 | XP_008250695.1 | 786 | 139 | |||
More Information
Set Membership: |
|

LOC100008606
Genomic Map