Assay Details
Gene Symbol: LOC100008640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329072.1 | NP_001316001.1 | 4 - 5 | 573 | 63 | |
XM_017347412.1 | XP_017202901.1 | 5 - 6 | 1264 | 63 | ||
XM_017347413.1 | XP_017202902.1 | 5 - 6 | 1084 | 63 | ||
XM_017347414.1 | XP_017202903.1 | 5 - 6 | 653 | 63 | ||
XM_017347416.1 | XP_017202905.1 | 5 - 6 | 1268 | 63 | ||
GenBank mRNA | AB231853.1 | - | 2 - 3 | 272 | 63 | |
KT852940.1 | - | 4 - 5 | 573 | 63 |
Gene Symbol: LOC100346814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017347417.1 | XP_017202906.1 | 4 - 5 | 544 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
