Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713474.3 | XP_002713520.1 | 2 - 3 | 516 | 60 | |
| XM_008261167.2 | XP_008259389.1 | 2 - 3 | 574 | 60 | ||
| GenBank mRNA | AF000312.1 | - | 1 - 2 | 30 | 60 | |
More Information
Set Membership: |
|

AQP4
Genomic Map