Assay Details
| Gene Symbol: TUBA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195806.1 | NP_001182735.1 | 4 | 1437 | 119 | |
| GenBank mRNA | AF011561.1 | - | 1 | 44 | 118 | |
| Gene Symbol: LOC100348075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714094.3 | XP_002714140.1 | 1 | 1443 | 119 | |
More Information
Set Membership: |
|

TUBA1B
Genomic Map