Assay Details
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195691.1 | NP_001182620.1 | 723 | 66 | ||
XM_008250271.1 | XP_008248493.1 | 796 | 66 | |||
XM_008250272.2 | XP_008248494.1 | 787 | 66 | |||
XM_008250273.2 | XP_008248495.1 | 715 | 66 | |||
GenBank mRNA | AF020290.1 | - | 59 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
