Assay Details
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195690.1 | NP_001182619.1 | 4 - 5 | 478 | 74 | |
GenBank mRNA | AF069715.1 | - | 3 - 4 | 326 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
