Assay Details
| Gene Symbol: TUBA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195806.1 | NP_001182735.1 | 2 - 3 | 69 | ||
| GenBank mRNA | AF200353.1 | - | 1 | 74 | 69 | |
| Gene Symbol: LOC100346652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718586.3 | XP_002718632.1 | 1 | 69 | ||
| Gene Symbol: LOC100348075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714094.3 | XP_002714140.1 | 1 | 69 | ||
| Gene Symbol: LOC100357567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256416.2 | XP_008254638.1 | 2 - 3 | 69 | ||
More Information
Set Membership: |
|

TUBA1B
Genomic Map