Assay Details
| Gene Symbol: TUBA1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195806.1 | NP_001182735.1 | 3 - 4 | 468 | 79 | |
| GenBank mRNA | AF200353.1 | - | 1 | 226 | 79 | |
| Gene Symbol: LOC100340483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711101.3 | XP_002711147.1 | 3 - 4 | 477 | 79 | |
| Gene Symbol: LOC100348075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714094.3 | XP_002714140.1 | 1 | 474 | 79 | |
| Gene Symbol: LOC100357567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256416.2 | XP_008254638.1 | 3 - 4 | 79 | ||
More Information
Set Membership: |
|

TUBA1B
Genomic Map