Assay Details
| Gene Symbol: HSD17B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345722.1 | XP_017201211.1 | 3 - 4 | 97 | ||
| GenBank mRNA | AF265660.1 | - | 333 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSD17B7| Gene Symbol: HSD17B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017345722.1 | XP_017201211.1 | 3 - 4 | 97 | ||
| GenBank mRNA | AF265660.1 | - | 333 | 97 | ||
Set Membership: |
|