Assay Details
Gene Symbol: LOC100008830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708253.3 | XP_002708299.1 | 6 - 7 | 906 | 66 | |
XM_017342293.1 | XP_017197782.1 | 3 - 4 | 514 | 66 | ||
GenBank mRNA | AF458078.1 | - | 2 - 3 | 365 | 66 | |
EF472893.1 | - | 4 - 5 | 656 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
