Assay Details
| Gene Symbol: PITX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347345.1 | XP_017202834.1 | 2 - 3 | 999 | 82 | |
| XM_017347346.1 | XP_017202835.1 | 3 - 4 | 555 | 82 | ||
| GenBank mRNA | AF474182.1 | - | 1 - 2 | 167 | 82 | |
More Information
Set Membership: |
|

PITX2
Genomic Map