Assay Details
| Gene Symbol: HMOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711415.3 | XP_002711461.1 | 2 - 3 | 294 | 74 | |
| GenBank mRNA | AY421756.1 | - | 1 - 2 | 93 | 74 | |
More Information
Set Membership: |
|

HMOX1
Genomic Map