Assay Details
| Gene Symbol: CER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002707983.2 | XP_002708029.1 | 1 - 2 | 588 | 78 | |
| GenBank mRNA | AY570542.1 | - | 1 - 2 | 480 | 78 | |
More Information
Set Membership: |
|

CER1
Genomic Map