Assay Details
| Gene Symbol: CLNS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171027.1 | NP_001164498.1 | 1 - 2 | 174 | 70 | |
| XM_017347854.1 | XP_017203343.1 | 1 - 2 | 211 | 70 | ||
| GenBank mRNA | AF003907.1 | - | 1 - 2 | 125 | 70 | |
| D26076.1 | - | 1 - 2 | 173 | 70 | ||
More Information
Set Membership: |
|

CLNS1A
Genomic Map