Assay Details
| Gene Symbol: FKBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718166.3 | XP_002718212.1 | 4 - 5 | 494 | 119 | |
| GenBank mRNA | D82876.1 | - | 4 - 5 | 452 | 119 | |
| Gene Symbol: LOC100348481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343517.1 | XP_017199006.1 | 1 | 480 | 119 | |
More Information
Set Membership: |
|

FKBP3
Genomic Map