Assay Details
| Gene Symbol: CXCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008267697.2 | XP_008265919.1 | 3 - 4 | 333 | 74 | |
| GenBank mRNA | EU407474.1 | - | 2 - 3 | 219 | 74 | |
More Information
Set Membership: |
|

CXCL13
Genomic Map