Assay Details
| Gene Symbol: TNFSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081985.1 | NP_001075454.1 | 1 - 2 | 252 | 122 | |
| GenBank mRNA | AB003912.1 | - | 1 - 2 | 252 | 122 | |
More Information
Set Membership: |
|

TNFSF4
Genomic Map