Assay Details
Gene Symbol: CLCN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082068.1 | NP_001075537.1 | 9 - 10 | 1936 | 72 | |
XM_008272337.2 | XP_008270559.1 | 13 - 14 | 2613 | 72 | ||
XM_008272338.2 | XP_008270560.1 | 10 - 11 | 2285 | 72 | ||
XM_008272339.2 | XP_008270561.1 | 10 - 11 | 2177 | 72 | ||
GenBank mRNA | AF195523.1 | - | 9 - 10 | 1936 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
