Assay Details
| Gene Symbol: ATP1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082073.1 | NP_001075542.1 | 4 - 5 | 584 | 87 | |
| GenBank mRNA | AF204927.1 | - | 4 - 5 | 584 | 87 | |
More Information
Set Membership: |
|

ATP1B1
Genomic Map