Assay Details
Gene Symbol: ATP1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082091.1 | NP_001075560.1 | 4 - 5 | 547 | 71 | |
GenBank mRNA | AF302929.1 | - | 4 - 5 | 547 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
