Assay Details
| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082109.1 | NP_001075578.1 | 937 | 103 | ||
| GenBank mRNA | AF367428.1 | - | 937 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKACA| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082109.1 | NP_001075578.1 | 937 | 103 | ||
| GenBank mRNA | AF367428.1 | - | 937 | 103 | ||
Set Membership: |
|