Assay Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082143.1 | NP_001075612.1 | 3 - 4 | 444 | 87 | |
| GenBank mRNA | AY094175.1 | - | 3 - 4 | 444 | 87 | |
More Information
Set Membership: |
|

PEBP1
Genomic Map