Assay Details
| Gene Symbol: S100A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082163.1 | NP_001075632.1 | 1 - 2 | 132 | 93 | |
| GenBank mRNA | AY452731.1 | - | 1 - 2 | 132 | 93 | |
More Information
Set Membership: |
|

S100A10
Genomic Map