Assay Details
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082172.1 | NP_001075641.1 | 8 - 9 | 1098 | 65 | |
GenBank mRNA | AY618931.1 | - | 8 - 9 | 1098 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
