Assay Details
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082200.1 | NP_001075669.1 | 1 - 2 | 115 | 108 | |
GenBank mRNA | D21265.1 | - | 1 - 2 | 115 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
