Assay Details
| Gene Symbol: SFTPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082229.1 | NP_001075698.1 | 345 | 111 | ||
| XM_008274301.2 | XP_008272523.1 | 416 | 111 | |||
| XM_017350523.1 | XP_017206012.1 | 413 | 111 | |||
| XM_017350524.1 | XP_017206013.1 | 405 | 111 | |||
| GenBank mRNA | J03542.1 | - | 345 | 111 | ||
More Information
Set Membership: |
|

SFTPA1
Genomic Map