Assay Details
| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082230.1 | NP_001075699.1 | 1 - 2 | 116 | 84 | |
| GenBank mRNA | J03744.1 | - | 1 - 2 | 116 | 84 | |
More Information
Set Membership: |
|

PMP2
Genomic Map