Assay Details
Gene Symbol: AKR1C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082250.1 | NP_001075719.1 | 578 | 86 | ||
GenBank mRNA | L17006.1 | - | 578 | 86 |
Gene Symbol: PGER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287557.1 | NP_001274486.1 | 86 | |||
GenBank mRNA | AB854081.1 | - | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
