Assay Details
| Gene Symbol: UBE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082296.1 | NP_001075765.1 | 5 - 6 | 336 | 131 | |
| GenBank mRNA | M62387.1 | - | 5 - 6 | 336 | 131 | |
More Information
Set Membership: |
|

UBE2B
Genomic Map