Assay Details
| Gene Symbol: PPP1R3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082303.1 | NP_001075772.1 | 2 - 3 | 857 | 142 | |
| GenBank mRNA | M65109.1 | - | 2 - 3 | 857 | 142 | |
More Information
Set Membership: |
|

PPP1R3A
Genomic Map