Assay Details
| Gene Symbol: GHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082636.1 | NP_001076105.1 | 4 - 5 | 416 | 81 | |
| XM_008262046.2 | XP_008260268.1 | 6 - 7 | 604 | 81 | ||
| XM_008262047.2 | XP_008260269.1 | 5 - 6 | 551 | 81 | ||
| XM_008262048.2 | XP_008260270.1 | 5 - 6 | 459 | 81 | ||
| XM_008262049.2 | XP_008260271.1 | 4 - 5 | 386 | 81 | ||
| XM_008262050.2 | XP_008260272.1 | 4 - 5 | 406 | 81 | ||
| XM_008262052.2 | XP_008260274.2 | 4 - 5 | 302 | 81 | ||
| XM_017344508.1 | XP_017199997.1 | 6 - 7 | 600 | 81 | ||
| XM_017344510.1 | XP_017199999.1 | 5 - 6 | 455 | 81 | ||
| GenBank mRNA | AF015252.1 | - | 4 - 5 | 416 | 81 | |
More Information
Set Membership: |
|

GHR
Genomic Map