Assay Details
Gene Symbol: SLC3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082656.1 | NP_001076125.1 | 902 | 61 | ||
GenBank mRNA | AF515773.1 | - | 902 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
