Assay Details
Gene Symbol: CST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082706.1 | NP_001076175.1 | 1 - 2 | 283 | 93 | |
GenBank mRNA | AB009342.1 | - | 1 - 2 | 283 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
