Assay Details
| Gene Symbol: DKK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082737.2 | NP_001076206.1 | 1 - 2 | 388 | 72 | |
| GenBank mRNA | AY575211.1 | - | 1 - 2 | 389 | 72 | |
| JN966759.1 | - | 1 - 2 | 415 | 72 | ||
More Information
Set Membership: |
|

DKK1
Genomic Map