Assay Details
| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082768.1 | NP_001076237.1 | 380 | 59 | ||
| GenBank mRNA | AJ131361.1 | - | 380 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC10A1| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082768.1 | NP_001076237.1 | 380 | 59 | ||
| GenBank mRNA | AJ131361.1 | - | 380 | 59 | ||
Set Membership: |
|